Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Heparan Sulfate 3-O-Sulfotransferase 4/HS3ST4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6085
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6085 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB6085 Supplier R&D Systems Supplier No. MAB6085
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Heparan Sulfate 3-O-Sulfotransferase 4/HS3ST4 Monoclonal specifically detects Heparan Sulfate 3-O-Sulfotransferase 4/HS3ST4 in Human samples. It is validated for Immunohistochemistry, Immunoprecipitation.

Specifications

Antigen Heparan Sulfate 3-O-Sulfotransferase 4/HS3ST4
Applications Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone 712010
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 8-25 ug/mL, Immunoprecipitation 25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9Y661
Gene Alias 30ST4, 3-OST-4, 3OST4 heparan sulfate 3-O-sulfotransferase-4, EC 2.8.2, EC 2.8.2.23, h3-OST-4, heparan sulfate (glucosamine) 3-O-sulfotransferase 4, Heparan sulfate 3-O-sulfotransferase 4, Heparan sulfate D-glucosaminyl 3-O-sulfotransferase 4, heparan sulfate glucosamine 3-O-sulfotransferase 4
Gene Symbols HS3ST4
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Heparan Sulfate 3-O-Sulfotransferase 4/HS3ST4 Gly184-Lys456 Accession # Q9Y661
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9951
Test Specificity Detects human Heparan Sulfate 3-O-Sulfotransferase 4/HS3ST4 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human HS3ST3B1 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.