Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Heparan Sulfate 6-O-Sulfotransferase 1/HS6ST1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $468.00
Specifications
Antigen | Heparan Sulfate 6-O-Sulfotransferase 1/HS6ST1 |
---|---|
Clone | 584308 |
Concentration | LYOPH |
Dilution | Immunohistochemistry 8-25 ug/mL |
Applications | Immunohistochemistry |
Description
Heparan Sulfate 6-O-Sulfotransferase 1/HS6ST1 Monoclonal specifically detects Heparan Sulfate 6-O-Sulfotransferase 1/HS6ST1 in Human samples. It is validated for Immunohistochemistry.Specifications
Heparan Sulfate 6-O-Sulfotransferase 1/HS6ST1 | |
LYOPH | |
Immunohistochemistry | |
Unconjugated | |
Mouse | |
Human | |
O60243 | |
9394 | |
Chinese hamster ovary cell line CHO-derived recombinant human Heparan Sulfate 6-O-Sulfotransferase 1/HS6ST1 Pro38-Trp411 Accession # O60243 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Heparan Sulfate 6-O-Sulfotransferase 1/HS6ST1 in direct ELISAs. |
584308 | |
Immunohistochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
DKFZp547H098, EC 2.8.2, EC 2.8.2.-, FLJ25392, heparan sulfate 6-O-sulfotransferase 1, heparan-sulfate 6-sulfotransferase, HS6ST-1, HS6STheparan-sulfate 6-O-sulfotransferase 1, MGC116899, MGC116901 | |
HS6ST1 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title