Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Hepassocin/FGL1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1614
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1614 500 μg
MAB1614SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB1614 Supplier R&D Systems Supplier No. MAB1614
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Hepassocin/FGL1 Monoclonal specifically detects Hepassocin/FGL1 in Human samples. It is validated for Western Blot.

Specifications

Antigen Hepassocin/FGL1
Applications Western Blot
Classification Monoclonal
Clone 198401
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. BAB70690
Gene Alias FGL1, fibrinogen-like 1, fibrinogen-like protein 1, Fibrinogen-like-protein 1, FREP1, Hepassocin, hepatocellular carcinoma-related sequence, Hepatocyte-derived fibrinogen-related protein 1, HFREP-1, HFREP1MGC12455, HP-041, LFIRE1, LFIRE-1, Liver fibrinogen-related protein 1
Gene Symbols FGL1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Hepassocin Met1-Ile312 Accession # BAB70690
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2267
Test Specificity Detects human Hepassocin in direct ELISAs and Western blots. In direct ELISAs, weak cross-reactivity with rat Hepassocin is observed. In Western blots, 100% cross-reactivity with rat Hepassocin is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.