Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Hepcidin Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$214.00 - $526.00
Specifications
Antigen | HepcidinAntimicrobialPeptide |
---|---|
Clone | 1033A |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB8307SP
![]() |
R&D Systems
MAB8307SP |
25 μg |
Each for $214.00
|
|
|||||
MAB8307100
![]() |
R&D Systems
MAB8307100 |
100 μg |
Each for $526.00
|
|
|||||
Description
Hepcidin Antimicrobial Peptide Monoclonal specifically detects Hepcidin Antimicrobial Peptide in Human samples. It is validated for ELISA.Specifications
HepcidinAntimicrobialPeptide | |
ELISA | |
Monoclonal | |
Supernatant | |
Human | |
P81172 | |
57817 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
1033A | |
ELISA | |
Unconjugated | |
Rabbit | |
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
HAMP, HEPC, hepcidin antimicrobial peptide, HEPCPutative liver tumor regressor, HFE2B, HFE2Bhepcidin, LEAP1, LEAP-1, LEAP1PLTR, Liver-expressed antimicrobial peptide 1, PLTR | |
Synthetic peptide containing human Hepcidin Accession # P81172 | |
Primary | |
Reconstitute at 0.5mg/mL in sterile PBS. | |
Detects human Hepcidin in direct ELISAs |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title