Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Hepsin Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB4776
This item is not returnable.
View return policy
Description
Hepsin Monoclonal specifically detects Hepsin in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| Hepsin | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| P05981 | |
| HPN | |
| Mouse myeloma cell line NS0-derived recombinant human Hepsin Ile163-Leu417 Accession # P05981 | |
| 100 μg | |
| Primary | |
| Detects human Hepsin in direct ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| 506220 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.4.21, EC 3.4.21.106, hepsin, HPN, serine protease hepsin, serine protease hepsin catalytic chain, TMPRSS1, TMPRSS1serine protease hepsin non-catalytic chain, Transmembrane protease serine 1, transmembrane protease, serine 1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3249 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction