Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human Herpes Simplex Virus 1 Antibody (IgM) ELISA Kit (Colorimetric)
SDP

Catalog No. NB600531KT Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600531KT 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600531KT Supplier Novus Biologicals™ Supplier No. NBP2600531KIT
Add to Cart
Add to Cart

CCL22 Colorimetric ELISA Kit is useful in the study of Cytokine, chemokine, Chemotaxis

The Human Herpes Simplex Virus 1 Antibody (IgM) ELISA Kit (Colorimetric) measures human Herpes Simplex Virus 1 Antibody (IgM) in serum, plasma.

Specifications

Assay Range Qualitative
Product Type ELISA Kit (Colorimetric)
Sample Type Plasma, Serum
Specificity Detection of human HSV antibody (IgM)
Target Species Human
For Use With (Application) ELISA
Kit Contents Coated assay plate, Negative Control, Positive Control, Sample Diluents, HRP- conjugate, Wash Buffer (30X concentrate), Substrate A, Substrate B, Stop Solution, Adhesive Strip (For 96 wells), Instruction manual
Quantity 1 Kit
Regulatory Status RUO
Research Discipline Cell Biology
Sample Volume 50 to 100 μL
Storage Requirements Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date.
Target Herpes Simplex Virus 1 Antibody (IgM)
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.