Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human HES-4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB3600
This item is not returnable.
View return policy
Description
HES-4 Monoclonal specifically detects HES-4 in Human samples. It is validated for Western Blot.Specifications
HES-4 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
Q9HCC6 | |
HES4 | |
E. coli-derived recombinant human HES-4 Val94-Arg221 Accession # Q9HCC6 | |
100 μg | |
Primary | |
Detects human HES-4 in Western blots. | |
Human | |
Purified |
Western Blot | |
455237 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
bHLH factor Hes4, BHLHB42, bHLHb42transcription factor HES-4, Class B basic helix-loop-helix protein 42, Hairy and enhancer of split 4, hairy and enhancer of split 4 (Drosophila), HES4, hHES4 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
57801 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction