Learn More
Human Hexokinase 1/2 Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | Hexokinase 1/2 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000 dilution, Immunohistochemistry 1:200 dilution |
| Formulation | Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. See Certificate of Analysis for details. with Sodium Azide |
| Gene Accession No. | P19367 & P52789 |
| Host Species | Rabbit |
| Immunogen | E. coli-derived recombinant human Hexokinase 1 and E. coli-derived recombinant human Hexokinase 2 Phe11-Ser917 and Phe11-Arg917 Accession # P19367 & P52789 |
| Purification Method | Immunogen affinity purified |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.