Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human HGF Activator Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $549.50
Specifications
Antigen | HGF Activator |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunohistochemistry, Immunoprecipitation |
Classification | Polyclonal |
Description
HGF Activator Polyclonal specifically detects HGF Activator in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.Specifications
HGF Activator | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL | |
Polyclonal | |
Goat | |
Human | |
HGF activator, HGFA, Hgfac | |
HGFAC | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunohistochemistry, Immunoprecipitation | |
Unconjugated | |
RUO | |
Q04756.1 | |
3083 | |
Mouse myeloma cell line NS0-derived recombinant human HGF Activator Gln36-Ser655 Accession # Q04756.1 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human HGF Activator in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 5% cross-reactivity with recombinant mouse HGF Activator is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title