Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human HGF Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF294NA
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF294NA 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF294NA Supplier R&D Systems Supplier No. AF294NA
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

HGF Polyclonal specifically detects HGF in Human samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.

Specifications

Antigen HGF
Applications Western Blot, Immunohistochemistry, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 0.01-.06 ug/mL
Gene Accession No. P14210
Gene Alias deafness, autosomal recessive 39, DFNB39, EC 3.4.21, EC 3.4.21.7, fibroblast-derived tumor cytotoxic factor, F-TCF, hepatocyte growth factor (hepapoietin A; scatter factor), Hepatopoeitin-A, Hepatopoietin A, HGFB, HPTA, HPTAhepatocyte growth factor, lung fibroblast-derived mitogen, Scatter factor, SF, SFhepatopoeitin-A
Gene Symbols HGF
Host Species Goat
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human HGF Accession # P14210
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3082
Test Specificity Detects human HGF in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant mouse HGF is observed. In Western blots, no cross-reactivity with recombinant canine HGF is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.