Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gibco™ Human HGF Recombinant Protein

Click to view available options
Quantity:
10 μg
Description
Human HGF Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
Recombinant Human Hepatocyte Growth Factor (HGF) is a bioactive protein intended for use in cell culture applications. HGF plays a role in positive regulation of cell proliferation.
High purity—no interference from other proteins or contaminants
High biological activity—more results with less protein
Proven compatibility—Gibco™ proteins bioassayed with Gibco™ media
High purity recombinant bioactive protein
Recombinant Human Hepatocyte Growth Factor is one of our more than 250 available Gibco™ recombinant proteins. To help ensure Gibco™ growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein.
High biological activity
In-house activity testing of Gibco™ Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, antiviral, and protease assays. Recombinant Human Hepatocyte Growth Factor (HGF) specific activity is determined by the dose dependent proliferation of monkey 4MBr-5 cells.
Proven compatibility
Gibco™ proteins are bioassayed with Gibco™ media. Since 1962, Gibco™ has been the consistent provider of high quality media, reagents, and sera for reliable cell culture.
See our selection of recombinant Gibco™ growth factors.
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.
High purity recombinant bioactive protein
Recombinant Human Hepatocyte Growth Factor is one of our more than 250 available Gibco™ recombinant proteins. To help ensure Gibco™ growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein.
High biological activity
In-house activity testing of Gibco™ Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, antiviral, and protease assays. Recombinant Human Hepatocyte Growth Factor (HGF) specific activity is determined by the dose dependent proliferation of monkey 4MBr-5 cells.
Proven compatibility
Gibco™ proteins are bioassayed with Gibco™ media. Since 1962, Gibco™ has been the consistent provider of high quality media, reagents, and sera for reliable cell culture.
See our selection of recombinant Gibco™ growth factors.
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.

Specifications
Specifications
For Use With (Application) | Cell Culture |
Gene ID (Entrez) | 3082 |
Molecular Weight (g/mol) | 80 kDa |
Purification Method | Sequential Chromatography |
Quantity | 10 μg |
Gene Alias | HGF |
Species | Human |
Content And Storage | Store in freezer (-5 to -30°C). |
Endotoxin Level | < 0.1 ng/μg |
Shipping Condition | Approved for shipment on Wet or Dry Ice |
Show More |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction