Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human HGFR/c-MET Antibody, Novus Biologicals™

Rabbit Monoclonal Antibody
$177.10 - $433.50
Specifications
Antigen | HGFR/c-MET |
---|---|
Clone | 2504B |
Dilution | Immunocytochemistry 3-25 ug/mL |
Applications | Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB11320100
![]() |
R&D Systems
MAB11320100 |
100 μg |
Each for $433.50
|
|
|||||
MAB11320SP
![]() |
R&D Systems
MAB11320SP |
25 μg |
Each for $177.10
|
|
|||||
Description
HGFR/c-MET Monoclonal antibody specifically detects HGFR/c-MET in Human samples. It is validated for ImmunocytochemistrySpecifications
HGFR/c-MET | |
Immunocytochemistry 3-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
AUTS9, c-Met, cMET, EC 2.7.10, EC 2.7.10.1, hepatocyte growth factor receptor, HGF R, HGF receptor, HGF/SF receptor, HGFR, MET, Met (c-Met), met proto-oncogene (hepatocyte growth factor receptor), met proto-oncogene tyrosine kinase, oncogene MET, Proto-oncogene c-Met, RCCP2, Scatter factor receptor, SF receptor, Tyrosine-protein kinase Met | |
Synthetic peptide HGFR_Y1234, Accession # P08581 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
2504B | |
Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human | |
P08581 | |
4233 | |
IgG | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title