Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human HIF-2 alpha/EPAS1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB2886
This item is not returnable.
View return policy
Description
HIF-2 alpha/EPAS1 Monoclonal specifically detects HIF-2 alpha/EPAS1 in Human samples. It is validated for Western Blot.Specifications
HIF-2 alpha/EPAS1 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
Q99814 | |
EPAS1 | |
E. coli-derived recombinant human HIF-2 alpha/EPAS1 Ser543-Thr870 Accession # Q99814 | |
100 μg | |
Primary | |
Detects human HIF-2 alpha/EPAS1 in Western blots. | |
Human | |
Purified |
Western Blot | |
318404 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Basic-helix-loop-helix-PAS protein MOP2, BHLHE73, Class E basic helix-loop-helix protein 73, ECYT4, endothelial PAS domain protein 1, endothelial PAS domain-containing protein 1, EPAS1, EPAS-1, HIF 2A, HIF-1-alpha-like factor, HIF-1alpha-like factor, HIF2 alpha, HIF-2 alpha, HIF2A, hif2a angiogenesis, HIF-2-alpha, HIF2-alpha, HLF, hypoxia-inducible factor 2 alpha, Hypoxia-inducible factor 2-alpha, Member of PAS protein 2, MOP2, PAS domain-containing protein 2, PASD2 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2034 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction