Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human HIN-1/SCGB3A1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2790
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2790 100 μg
AF2790SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF2790 Supplier R&D Systems Supplier No. AF2790
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

HIN-1/SCGB3A1 Polyclonal specifically detects HIN-1/SCGB3A1 in Human samples. It is validated for Western Blot.

Specifications

Antigen HIN-1/SCGB3A1
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Gene Accession No. Q96QR1
Gene Alias cytokine high in normal-1, Cytokine HIN-1, High in normal 1, HIN1, HIN-1, HIN1Pneumo secretory protein 2, LU105, PnSP-2, PNSP2, PnSP-2Uteroglobin-related protein 2, SCGB3A1, secretoglobin family 3A member 1, secretoglobin, family 3A, member 1, UGRP2, UGRP2MGC87867
Gene Symbols SCGB3A1
Host Species Goat
Immunogen E. coli-derived recombinant human HIN-1/SCGB3A1 Phe21-Gly104 Accession # Q96QR1
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92304
Test Specificity Detects human HIN-1/SCGB3A1 in direct ELISAs and Western blots. In Western blots, approximately 10% cross-reactivity with recombinant mouse HIN-1 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.