Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human HIPK1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF5617
This item is not returnable.
View return policy
Description
HIPK1 Polyclonal specifically detects HIPK1 in Human samples. It is validated for Western Blot.Specifications
HIPK1 | |
Polyclonal | |
Unconjugated | |
Q86Z02 | |
HIPK1 | |
E. coli-derived recombinant human HIPK1 Gln701-Ser900 Accession # Q86Z02 | |
100 μg | |
Primary | |
Detects endogenous human HIPK1 in Western blots. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
EC 2.7.11.1, homeodomain interacting protein kinase 1, homeodomain interacting protein kinase 1-like protein, homeodomain-interacting protein kinase 1, KIAA0630MGC26642, MGC33446, MGC33548, Myak, Nbak2, nuclear body associated kinase 2b | |
Goat | |
Affinity Purified | |
RUO | |
204851 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction