Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human HIPK2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $491.00
Specifications
Antigen | HIPK2 |
---|---|
Clone | 493918 |
Dilution | Immunocytochemistry 5-25 ug/mL |
Applications | Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB9307SP
![]() |
R&D Systems
MAB9307SP |
25 μg |
Each for $200.50
|
|
|||||
MAB9307100
![]() |
R&D Systems
MAB9307100 |
100 μg |
Each for $491.00
|
|
|||||
Description
HIPK2 Monoclonal specifically detects HIPK2 in Human samples. It is validated for Immunocytochemistry.Specifications
HIPK2 | |
Immunocytochemistry 5-25 ug/mL | |
Monoclonal | |
Mouse | |
Human | |
Q9H2X6 | |
28996 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
493918 | |
Immunocytochemistry | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
DKFZp686K02111, EC 2.7.11.1, FLJ23711, hHIPk2, homeodomain interacting protein kinase 2, homeodomain-interacting protein kinase 2, PRO0593, Stank | |
E. coli-derived recombinant human HIPK2 Met765-Ser924 Accession # Q9H2X6 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human HIPK2 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title