Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Histamine H1 R Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB4726U100UG
This item is not returnable.
View return policy
Description
Histamine H1R Monoclonal specifically detects Histamine H1R in Human samples. It is validated for Flow Cytometry.Specifications
Histamine H1R | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
AAH60802 | |
HRH1 | |
NS0 mouse myeloma cell line transfected with human Histamine H1 R Met1-Ser487 Accession # AAH60802 | |
RUO | |
3269 | |
Human | |
IgG1 |
Flow Cytometry | |
480054 | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
H1R, H1-R, hisH1, Histamine H1R, histamine receptor H1, HRH1, subclass H1 | |
Mouse | |
100 μg | |
Primary | |
Detects human Histamine H1 R. Stains human Histamine H1 R transfectants but not irrelevant transfectants. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction