Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Histamine H1 R Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB4726SP
This item is not returnable.
View return policy
Description
Histamine H1R Monoclonal specifically detects Histamine H1R in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
Histamine H1R | |
Monoclonal | |
LYOPH | |
Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
AAH60802 | |
HRH1 | |
NS0 mouse myeloma cell line transfected with human Histamine H1 R Met1-Ser487 Accession # AAH60802 | |
25 μg | |
Primary | |
Detects human Histamine H1 R. Stains human Histamine H1 R transfectants but not irrelevant transfectants. | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
480054 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
H1R, H1-R, hisH1, Histamine H1R, histamine receptor H1, HRH1, subclass H1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3269 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction