Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Histamine H1 R Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB4726
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4726 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB4726 Supplier R&D Systems Supplier No. MAB4726
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Histamine H1R Monoclonal specifically detects Histamine H1R in Human samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen Histamine H1R
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 480054
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. AAH60802
Gene Alias H1R, H1-R, hisH1, Histamine H1R, histamine receptor H1, HRH1, subclass H1
Gene Symbols HRH1
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human Histamine H1 R Met1-Ser487 Accession # AAH60802
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3269
Test Specificity Detects human Histamine H1 R. Stains human Histamine H1 R transfectants but not irrelevant transfectants.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.