Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Histamine H3 R Alexa Fluor™ 350-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB10200U10
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB10200U10 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB10200U10 Supplier R&D Systems Supplier No. FAB10200U100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Histamine H3R Monoclonal specifically detects Histamine H3R in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Histamine H3R
Applications Flow Cytometry
Classification Monoclonal
Clone 1008944
Conjugate Alexa Fluor 350
Dilution Flow Cytometry 0.25-1 μg/106 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. Q9Y5N1
Gene Alias GPCR97, GPCR97G protein-coupled receptor 97, G-protein coupled receptor 97, H3R, HH3R, histamine H3 receptor, Histamine H3R, histamine receptor H3, HRH3
Host Species Mouse
Immunogen Synthetic peptide containing human Histamine H3 R, Accession # Q9Y5N1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 11255
Test Specificity Detects human Histamine H3 R in direct ELISAs.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Supernatant
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.