Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Histamine H3 R Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $456.00
Specifications
Antigen | Histamine H3R |
---|---|
Clone | 1008944 |
Dilution | Flow Cytometry 0.25 μg/106 cells, CyTOF-ready |
Applications | Flow Cytometry, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB10200SP
![]() |
R&D Systems
MAB10200SP |
25 μg |
Each for $200.50
|
|
|||||
MAB10200100
![]() |
R&D Systems
MAB10200100 |
100 μg |
Each for $456.00
|
|
|||||
Description
Histamine H3R Monoclonal specifically detects Histamine H3R in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
Histamine H3R | |
Flow Cytometry 0.25 μg/106 cells, CyTOF-ready | |
Monoclonal | |
Supernatant | |
Human | |
Q9Y5N1 | |
11255 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
1008944 | |
Flow Cytometry, CyTOF | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
GPCR97, GPCR97G protein-coupled receptor 97, G-protein coupled receptor 97, H3R, HH3R, histamine H3 receptor, Histamine H3R, histamine receptor H3, HRH3 | |
Synthetic peptide containing human Histamine H3 R, Accession # Q9Y5N1 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Histamine H3 R in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title