Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Histamine N-Methyltransferase/HNMT Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF7637SP
This item is not returnable.
View return policy
Description
Histamine N-Methyltransferase/HNMT Polyclonal specifically detects Histamine N-Methyltransferase/HNMT in Human samples. It is validated for Western Blot.Specifications
Histamine N-Methyltransferase/HNMT | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 2.1.1.8, histamine N-methyltransferase, Histamine NMethyltransferase, HMT, HNMT, HNMT-S1, HNMT-S2 | |
Sheep | |
Affinity Purified | |
RUO | |
3176 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 0.5 ug/mL | |
P50135 | |
HNMT | |
E. coli-derived recombinant human Histamine N-Methyltransferase/HNMT Ser7-Ala292 Accession # P50135 | |
25 μg | |
Primary | |
Detects human Histamine N-Methyltransferase/HNMT in direct ELISAs and Western blots. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction