Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Histone Deacetylase 4/HDAC4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $503.00
Specifications
| Antigen | Histone Deacetylase 4/HDAC4 |
|---|---|
| Clone | 713802 |
| Dilution | Immunohistochemistry 5-25 ug/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Conjugate | Quantity | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Conjugate | Quantity | Price | Quantity & Availability | ||||
MAB6205SP
![]() |
R&D Systems
MAB6205SP |
Unconjugated | 25 μg |
Each for $200.50
|
|
||||
MAB6205100
![]() |
R&D Systems
MAB6205100 |
Unconjugated | 100 μg |
Each for $503.00
|
|
||||
Description
Histone Deacetylase 4/HDAC4 Monoclonal specifically detects Histone Deacetylase 4/HDAC4 in Human samples. It is validated for Immunohistochemistry.Specifications
| Histone Deacetylase 4/HDAC4 | |
| Immunohistochemistry 5-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| HA6116, HD4EC 3.5.1.98, HDAC4, HDAC-A, HDACABDMR, histone deacetylase 4, histone deacetylase A, KIAA0288AHO3 | |
| E. coli-derived recombinant human Histone Deacetylase 4/HDAC4 Met1-Glu68 Accession # P56524 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Histone Deacetylase 4/HDAC4 in direct ELISAs. |
| 713802 | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P56524 | |
| 9759 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title