Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Histone H2AX Alexa Fluor 700-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems IC2288N100UG
This item is not returnable.
View return policy
Description
Histone H2AX Monoclonal specifically detects Histone H2AX in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
Histone H2AX | |
Monoclonal | |
Alexa Fluor 700 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
H2A.X, H2A/X, H2AFX | |
Phospho-peptide containing human Histone H2AX S139 site Accession # P16104 | |
100 μg | |
3014 | |
Human | |
Supernatant |
Flow Cytometry | |
2207D | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
P16104 | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
Primary | |
Detects human Histone H2AX when phosphorylated at S139 in Western blots. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction