Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human HLA-C Alexa Fluor™ 700-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB10394N10
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB10394N10 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB10394N10 Supplier R&D Systems Supplier No. FAB10394N100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

HLA C Monoclonal specifically detects HLA C in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen HLA C
Applications Flow Cytometry
Classification Monoclonal
Clone DT-9
Conjugate Alexa Fluor 700
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. P10321
Gene Alias C alpha chain, D6S204, FLJ27082, HLA class I histocompatibility antigen, Cw-1 alpha chain, HLAC, HLA-Cw, HLA-JY3, HLC-C, human leukocyte antigen-C alpha chain, leucocyte antigen C, leucocyte antigen Cw, leukocyte antigen-C alpha chain, major histocompatibility antigen HLA-C, major histocompatibility complex, class I, C, MHC class I antigen Cw*1, MHC class I antigen Cw*12, MHC class I antigen Cw*14, MHC class I antigen Cw*15, MHC class I antigen Cw*16, MHC class I antigen Cw*17, MHC class I antigen Cw*2, MHC class I antigen Cw*3, MHC class I antigen Cw*4, MHC class I antigen Cw*5, MHC class I antigen Cw*6, MHC class I antigen Cw*7, MHC class I antigen Cw*8, MHC class I antigen heavy chain HLA-C, PSORS1
Host Species Mouse
Immunogen Purified tamarin MHC class I molecules (using anti-class I antibody BB7.7), Accession # P10321
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3107
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.