Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human HLA Class I APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB7098A
This item is not returnable.
View return policy
Description
HLA Class I Monoclonal specifically detects HLA Class I in Human samples. It is validated for Flow Cytometry.Specifications
HLA Class I | |
Monoclonal | |
APC | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
HLA-A | |
Membranes from human tonsillar lymphocytes | |
100 Tests | |
Primary | |
Detects the human major histocompatibility complex (MHC) class I, HLA-A, B, and C. Recognizes a non-polymorphic epitope shared among products of the HLA-A, B, and C loci and immunoprecipitates both the HLA molecule and beta 2-Microglobulin. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2a |
Flow Cytometry | |
W6/32 | |
Flow Cytometry 10 uL/10^6 cells | |
A-10 alpha chain, FLJ26655, HLA class I histocompatibility antigen, A-1 alpha chain, HLA class I histocompatibility antigen, A-28 alpha chain, HLA class I histocompatibility antigen, A-9 alpha chain, HLAA, HLA-A, HLA-ABC, major histocompatibility complex, class I, A, MHC class I antigen A*1, MHC class I antigen A*11, MHC class I antigen A*80 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3105 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction