Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human HLA-DR PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
$245.00 - $502.50
Specifications
Antigen | HLA-DR |
---|---|
Clone | L203 |
Dilution | Flow Cytometry 10 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB4869P025
![]() |
R&D Systems
FAB4869P025 |
25 Tests |
Each for $245.00
|
|
|||||
FAB4869P100
![]() |
R&D Systems
FAB4869P100 |
100 Tests |
Each for $502.50
|
|
|||||
Description
HLA-DR Monoclonal specifically detects HLA-DR in Human samples. It is validated for Flow Cytometry.Specifications
HLA-DR | |
Flow Cytometry 10 uL/10^6 cells | |
Monoclonal | |
Purified | |
RUO | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
FLJ51114, histocompatibility antigen HLA-DR alpha, HLA class II histocompatibility antigen, DR alpha chain, HLADR, HLA-DRA, HLA-DRA1, major histocompatibility complex, class II, DR alpha, MHC cell surface glycoprotein, MHC class II antigen DRA, MHC Class II DR, MLRW | |
HLA-DRA | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Detects human HLA-DR. |
L203 | |
Flow Cytometry | |
R-PE | |
Mouse | |
Human | |
P01903 | |
3122 | |
RPMI 8866 human lymphoblastoid cells Accession # P01903 | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title