Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human HM74A/PUMA-G/GPR109A Antibody, R&D Systems™
GREENER_CHOICE

Rat Monoclonal Antibody

$188.28 - $449.33

Specifications

Antigen HM74A/PUMA-G/GPR109A/NIACR1
Clone 245115
Dilution Immunohistochemistry 5-25 ug/mL
Applications Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB27601SP
GREENER_CHOICE
View Documents
R&D Systems
MAB27601SP
25 μg
Each for $188.28
Only null left
Add to Cart
 
MAB27601100
GREENER_CHOICE
View Documents
R&D Systems
MAB27601100
100 μg
Each for $449.33
Only null left
Add to Cart
 
Description

Description

HM74A/PUMA-G/GPR109A/NIACR1 Monoclonal specifically detects HM74A/PUMA-G/GPR109A/NIACR1 in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

HM74A/PUMA-G/GPR109A/NIACR1
Immunohistochemistry 5-25 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
GPR109A, HCA2, HCAR2, hydroxycarboxylic acid receptor 2, Niacr1, Nicotinic Acid Receptor, PUMAG
NS0 mouse myeloma cell line transfected with human HM74A/PUMA-G/GPR109A, Met1-Pro363, Accession # Q8TDS4
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human HM74A/PUMA-G/GPR109A in direct ELISAs.
245115
Immunohistochemistry
Unconjugated
Rat
Human
Q8TDS4
338442
IgG2b
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.