Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human HM74A/PUMA-G/GPR109A APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB2760A
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB2760A 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB2760A Supplier R&D Systems Supplier No. FAB2760A
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

HM74A/PUMA-G/GPR109A/NIACR1 Monoclonal specifically detects HM74A/PUMA-G/GPR109A/NIACR1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen HM74A/PUMA-G/GPR109A/NIACR1
Applications Flow Cytometry
Classification Monoclonal
Clone 245106
Conjugate APC
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q8TDS4
Gene Alias GPR109A, HCA2, HCAR2, hydroxycarboxylic acid receptor 2, Niacr1, Nicotinic Acid Receptor, PUMAG
Gene Symbols HCAR2
Host Species Rat
Immunogen NS0 mouse myeloma cell line transfected with human HM74A/PUMA-G/GPR109A Met1-Pro363 Accession # Q8TDS4
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 338442
Test Specificity Detects human HM74A/PUMA-G/GPR109A. Stains human HM74A/PUMA-G/GPR109A-transfected cells but not irrelevant transfectants. Cross-reactivity with HM74 (GPR109B) was not tested.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.