Learn More
Human HMGB1/HMG-1 Alexa Fluor™ 350-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | HMGB1/HMG-1 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 115603 |
| Concentration | 0.2 mg/mL |
| Conjugate | Alexa Fluor 350 |
| Dilution | Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells |
| Formulation | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide |
| Gene Accession No. | P09429 |
| Gene Alias | Amphoterin, high mobility group box 1, High mobility group protein 1, high mobility group protein B1, high-mobility group (nonhistone chromosomal) protein 1, high-mobility group box 1, HMG1, HMG-1, HMG1DKFZp686A04236, HMG3, SBP-1, Sulfoglucuronyl carbohydrate binding protein |
| Show More |
For Research Use Only (RUO)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.