Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human HMGB1/HMG-1 PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC1690P
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC1690P 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC1690P Supplier R&D Systems Supplier No. IC1690P
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Applications:

Chromatin Immunoprecipitation, Flow Cytometry, Immunocytochemistry, Western Blotting

Specifications

Antigen HMGB1/HMG-1
Applications Flow Cytometry
Classification Monoclonal
Clone 115603
Conjugate R-PE
Dilution Intracellular Staining by Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. P09429
Gene Alias Amphoterin, high mobility group box 1, High mobility group protein 1, high mobility group protein B1, high-mobility group (nonhistone chromosomal) protein 1, high-mobility group box 1, HMG1, HMG-1, HMG1DKFZp686A04236, HMG3, SBP-1, Sulfoglucuronyl carbohydrate binding protein
Gene Symbols HMGB1
Host Species Mouse
Immunogen E. coli-derived recombinant human HMGB1/HMG-1 Gly2-Glu215 (predicted) Accession # P09429
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3146
Test Specificity Detects human HMGB1/HMG-1 in direct ELISAs and Western blots.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.