Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human HNF-3 alpha/FoxA1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6778
This item is not returnable.
View return policy
Description
HNF-3 alpha/FoxA1 Monoclonal specifically detects HNF-3 alpha/FoxA1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.Specifications
HNF-3 alpha/FoxA1 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 1-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
P55317 | |
FOXA1 | |
E. coli-derived recombinant human HNF-3 alpha/FoxA1 Met253-Ser472 Accession # P55317 | |
100 μg | |
Primary | |
Detects human HNF-3 alpha/FoxA1 in direct ELISAs and Western blots. | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
654126 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
forkhead box A1, Forkhead box protein A1, FoxA1, hepatocyte nuclear factor 3, alpha, hepatocyte nuclear factor 3-alpha, HNF3 alpha, HNF-3A, HNF-3-alpha, HNF3ATCF-3A, MGC33105, TCF3A, Transcription factor 3A | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3169 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction