Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human HNF-3 beta/FoxA2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2400
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2400 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF2400 Supplier R&D Systems Supplier No. AF2400
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 48 publications

HNF-3 beta/FoxA2 Polyclonal specifically detects HNF-3 beta/FoxA2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Chromatin Immunoprecipitation (ChIP), Immunocytochemistry.

Specifications

Antigen HNF-3 beta/FoxA2
Applications ChIP Assay, Immunocytochemistry, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 3-15 ug/mL, Chromatin Immunoprecipitation (ChIP) 5 ug/5 x 10^6 cells, Immunocytochemistry 5-15 ug/mL
Gene Accession No. Q9Y261
Gene Alias forkhead box A2, Forkhead box protein A2, FoxA2, hepatic nuclear factor-3-beta, hepatocyte nuclear factor 3, beta, hepatocyte nuclear factor 3-beta, HNF3 beta, HNF-3B, HNF-3-beta, HNF3BTCF-3B, MGC19807, TCF3B, Transcription factor 3B
Gene Symbols FOXA2
Host Species Goat
Immunogen E. coli-derived recombinant human HNF-3 beta/FoxA2 Met242-Ser457 Accession # Q9Y261
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3170
Test Specificity Detects human HNF-3 beta/FoxA2 in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.