Learn More
Human HNF-4 alpha/NR2A1 Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | HNF-4 alpha/NR2A1 |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | K9218 |
| Conjugate | Unconjugated |
| Dilution | Western Blot, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation |
| Formulation | A liquid formulation in physiologic saline with 0.1% NaN 3 . with 0.1% Sodium Azide |
| Gene Alias | FLJ39654, hepatic nuclear factor 4 alpha, hepatocyte nuclear factor 4, alpha, hepatocyte nuclear factor 4-alpha, HNF4 alpha, HNF4A, HNF4a7, HNF-4-alpha, HNF4alpha10/11/12, HNF4HNF4a8, MODY, MODY1, NR2A1, NR2A1HNF4a9, NR2A21, Nuclear receptor subfamily 2 group A member 1, TCF, TCF14, TCF-14, TCF14HNF4alpha, Transcription factor 14, Transcription factor HNF-4, transcription factor-14 |
| Gene Symbols | HNF4A |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.