Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human HNF-4 alpha/NR2A1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF4605
This item is not returnable.
View return policy
Description
HNF-4 alpha/NR2A1 Polyclonal specifically detects HNF-4 alpha/NR2A1 in Human samples. It is validated for Western Blot, Simple Western.Specifications
HNF-4 alpha/NR2A1 | |
Polyclonal | |
Unconjugated | |
P41235 | |
HNF4A | |
E. coli-derived recombinant human HNF-4 alpha/NR2A1 Val130-Ser330 Accession # P41235 | |
100 μg | |
Primary | |
Detects human HNF-4 alpha/NR2A1 in Western blots. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL, Simple Western 50 ug/mL | |
FLJ39654, hepatic nuclear factor 4 alpha, hepatocyte nuclear factor 4, alpha, hepatocyte nuclear factor 4-alpha, HNF4 alpha, HNF4A, HNF4a7, HNF-4-alpha, HNF4alpha10/11/12, HNF4HNF4a8, MODY, MODY1, NR2A1, NR2A1HNF4a9, NR2A21, Nuclear receptor subfamily 2 group A member 1, TCF, TCF14, TCF-14, TCF14HNF4alpha, Transcription factor 14, Transcription factor HNF-4, transcription factor-14 | |
Goat | |
Affinity Purified | |
RUO | |
3172 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction