Learn More
Human HNF-6/ONECUT1 Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | HNF-6/ONECUT1 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL |
| Gene Accession No. | Q9UBC0 |
| Gene Alias | HNF6, HNF-6A, HNF6Ahepatocyte nuclear factor 6, alpha, HNF6hepatocyte nuclear factor 6, HNF-6One cut domain family member 1, OC-1, one cut homeobox 1one cut domain, family member 1, ONECUT1 |
| Gene Symbols | ONECUT1 |
| Host Species | Sheep |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.