Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human HOIP/RNF31 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB8039
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8039 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB8039 Supplier R&D Systems Supplier No. MAB8039
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 2 publications

HOIP/RNF31 Monoclonal specifically detects HOIP/RNF31 in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.

Specifications

Antigen HOIP/RNF31
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 875227
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Simple Western 5 ug/mL, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q96EP0
Gene Alias EC 6.3.2.-, FLJ10111, FLJ23501, HOIP, MGC19975, ring finger protein 31, RNF31, ZIBRA, ZIBRAHOIL-1-interacting protein, Zinc in-between-RING-finger ubiquitin-associated domain protein
Gene Symbols RNF31
Host Species Mouse
Immunogen E. coli-derived recombinant human HOIP/RNF31 Arg970-Lys1072 Accession # Q96EP0
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55072
Test Specificity Detects human HOIP/RNF31 in direct ELISA and Western Blot.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.