Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Hormone-sensitive Lipase/HSL Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB7104SP
This item is not returnable.
View return policy
Description
Hormone-sensitive Lipase/HSL Monoclonal specifically detects Hormone-sensitive Lipase/HSL in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Hormone-sensitive Lipase/HSL | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL | |
Q05469 | |
LIPE | |
E. coli-derived recombinant human Hormone-sensitive Lipase/HSL Met302-Leu425 Accession # Q05469 | |
25 μg | |
Primary | |
Detects human Hormone-sensitive Lipase/HSL in direct ELISAs. | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
729923 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
hormone-sensitive lipase, Hormonesensitive Lipase, hormone-sensitive lipase testicular isoform, HSL, HSLEC 3.1.1.79, LHS, lipase, hormone-sensitive, LIPE | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3991 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction