Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Hormone-sensitive Lipase/HSL Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF7104SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF7104SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF7104SP Supplier R&D Systems Supplier No. AF7104SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Hormone-sensitive Lipase/HSL Polyclonal specifically detects Hormone-sensitive Lipase/HSL in Human samples. It is validated for Western Blot.

Specifications

Antigen Hormone-sensitive Lipase/HSL
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q05469
Gene Alias hormone-sensitive lipase, Hormonesensitive Lipase, hormone-sensitive lipase testicular isoform, HSL, HSLEC 3.1.1.79, LHS, lipase, hormone-sensitive, LIPE
Gene Symbols LIPE
Host Species Sheep
Immunogen E. coli-derived recombinant human Hormone-sensitive Lipase/HSL Met302-Leu425 Accession # Q05469
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3991
Test Specificity Detects human Hormone-sensitive Lipase/HSL in direct ELISAs and Western blots.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.