Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human HOXA1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF5014
This item is not returnable.
View return policy
Description
HOXA1 Polyclonal specifically detects HOXA1 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
HOXA1 | |
Polyclonal | |
Unconjugated | |
P49639 | |
HOXA1 | |
E. coli-derived recombinant human HOXA1 Met1-Asp118 Accession # P49639 | |
100 μg | |
Primary | |
Detects human HOXA1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human (rh) HOXB1, rhHOXB4, and rhHOXA9 is observed. | |
Human | |
IgG |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
BSAS, homeo box A1, homeobox A1, Homeobox protein Hox-1F, homeobox protein Hox-A1, Hox 1.6-like protein, HOX A1 homeodomain protein, HOX1, HOX1F, HOX1Fhomeobox 1F, lab-like protein, MGC45232 | |
Goat | |
Affinity Purified | |
RUO | |
3198 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction