Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human HOXB1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF6318
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF6318 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF6318 Supplier R&D Systems Supplier No. AF6318
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

HOXB1 Polyclonal specifically detects HOXB1 in Human samples. It is validated for Immunocytochemistry.

Specifications

Antigen HOXB1
Applications Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunocytochemistry 5-15 ug/mL
Gene Accession No. P14653
Gene Alias homeo box 2I, homeo box B1, homeobox B1, Homeobox protein Hox-2I, homeobox protein Hox-B1, HOX2, Hox-2.9, HOX2I, HOX2IMGC116844, MGC116843, MGC116845
Gene Symbols HOXB1
Host Species Sheep
Immunogen E. coli-derived recombinant human HOXB1 Met1-Thr192 Accession # P14653
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3211
Test Specificity Detects human HOXB1 in direct ELISAs. In direct ELISAs, less than 2% cross-reactivity with recombinant human (rh) HOXA1, rhHOXB4, and rhHOXA9 is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.