Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human HOXD10 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF10413SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF10413SP 25 μg
AF10413100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF10413SP Supplier R&D Systems Supplier No. AF10413SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Antibody

HOXD10 specifically detects HOXD10 in Human samples. It is validated for Immunocytochemistry.

Specifications

Antigen HOXD10
Applications Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P28358
Gene Alias homeo box 4D, homeo box D10, homeobox D10, Homeobox protein Hox-4D, Homeobox protein Hox-4E, homeobox protein Hox-D10, HOX4, Hox-4.4, HOX4D, HOX4DHOX4, HOX4E
Host Species Goat
Immunogen E. coli-derived recombinant human HOXD10, Arg153-Thr262, Accession # P28358
Purification Method Antigen Affinity-purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3236
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.