Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human HPRG Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1869
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1869 500 μg
MAB1869SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB1869 Supplier R&D Systems Supplier No. MAB1869
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

HPRG Monoclonal specifically detects HPRG in Human samples. It is validated for Western Blot.

Specifications

Antigen HPRG
Applications Western Blot
Classification Monoclonal
Clone 227901
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P04196
Gene Alias DKFZp779H1622, Histidine-proline-rich glycoprotein, histidine-rich glycoprotein, HPRGHRGP, HRG
Gene Symbols HRG
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human HPRG Val19-Lys525 Accession # P04196
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3273
Test Specificity Detects human HPRG in direct ELISAs and Western blots. In direct ELISAs, this antibody does not cross-react with recombinant human (rh) Cystatin B, C, D, E/M, S, A, SA, SN, rhFetuin A, B, rmHPRG, rhKininogen, or rhKininostatin.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.