Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human HS3ST3A1 Partial ORF (NP_006033, 308 a.a. - 406 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. p-7098344
Click to view available options
Quantity:
10 μg
25 μg

Used for AP, Array, ELISA, WB-Re

Heparan sulfate biosynthetic enzymes are key components in generating a myriad of distinct heparan sulfate fine structures that carry out multiple biologic activities. The enzyme encoded by this gene is a member of the heparan sulfate biosynthetic enzyme family. It is a type II integral membrane protein and possesses heparan sulfate glucosaminyl 3-O-sulfotransferase activity. The sulfotransferase domain of this enzyme is highly similar to the same domain of heparan sulfate D-glucosaminyl 3-O-sulfotransferase 3A1, and these two enzymes sulfate an identical disaccharide. This gene is widely expressed, with the most abundant expression in liver and placenta. [provided by RefSeq]

Sequence: LFVSGERLISDPAGELGRVQDFLGLKRIITDKHFYFNKTKGFPCLKKAEGSSRPHCLGKTKGRTHPEIDREVVRRLREFYRPFNLKFYQMTGHDFGWDG

Specifications

Accession Number NP_006033
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 9955
Molecular Weight (g/mol) 36.63kDa
Name HS3ST3A1 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen LFVSGERLISDPAGELGRVQDFLGLKRIITDKHFYFNKTKGFPCLKKAEGSSRPHCLGKTKGRTHPEIDREVVRRLREFYRPFNLKFYQMTGHDFGWDG
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias 30ST3A1/3OST3A1
Common Name HS3ST3A1
Gene Symbol HS3ST3A1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.