Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human HSP10/EPF Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB3298SP
This item is not returnable.
View return policy
Description
HSP10/EPF Monoclonal specifically detects HSP10/EPF in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
HSP10/EPF | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
P61604 | |
HSPE1 | |
E. coli-derived recombinant human HSP10/EPF Ala2-Asp102 Accession # P61604 | |
25 μg | |
Primary | |
Detects human HSP10/EPF in Western blots. | |
Human | |
Purified |
Western Blot, Immunocytochemistry | |
330629 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
10 kDa chaperonin, 10 kDa heat shock protein, mitochondrial, Chaperonin 10, Chaperonin 10 Homolog, CPN10HSP10, Early-pregnancy factor, EPF, GroES, heat shock 10kD protein 1 (chaperonin 10), heat shock 10kDa protein 1 (chaperonin 10), Hsp10, HSPE1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3336 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction