Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human HSP47 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $514.50
Specifications
Antigen | Hsp47 |
---|---|
Clone | 950811 |
Dilution | Western Blot 0.1 ug/mL, Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 5-25 ug/mL |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB91661SP
![]() |
R&D Systems
MAB91661SP |
25 μg |
Each for $200.50
|
|
|||||
MAB91661100
![]() |
R&D Systems
MAB91661100 |
100 μg |
Each for $514.50
|
|
|||||
Description
Hsp47 Monoclonal specifically detects Hsp47 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.Specifications
Hsp47 | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 5-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
(collagen binding protein 1), Arsenic-transactivated protein 3, AsTP3, BERF-1, CBP1, CBP2, CBP2RA-A47, Cell proliferation-inducing gene 14 protein, Collagen-binding protein, colligen, Colligin, colligin-1, colligin-2, gp46, HSP47serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), member 1, (collagen binding protein 1), member 2, member 2, (collagen-binding protein 2), OI10, PPROM, proliferation-inducing gene 14, RA-A47, rheumatoid arthritis antigen A-47,47 kDa heat shock protein, Rheumatoid arthritis-related antigen RA-A47, serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), serpin H1, serpin peptidase inhibitor, clade H (heat shock protein 47), member 1, SERPINH1, SERPINH2colligin | |
SERPINH1 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
950811 | |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Human | |
P50454 | |
871 | |
Chinese hamster ovary cell line CHO-derived recombinant human HSP47 Ala19-Asp412 Accession # P50454 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human HSP47 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with mouse HSP47 is observed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title