Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human HspA6 ELISA Kit (Colorimetric)
SDP

Catalog No. NBP262153 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP262153 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP262153 Supplier Novus Biologicals™ Supplier No. NBP262153
Add to Cart
Add to Cart

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.

The Human HspA6 ELISA Kit (Colorimetric) measures human HspA6 in cell lysates, serum, and tissue.

Specifications

Assay Range 0.156 - 10 ng/ml
Assay Sensitivity 62 pg/ml
Product Type ELISA Kit (Colorimetric)
Sample Type Cell Lysates, Serum, Tissue
For Use With (Application) ELISA
Gene ID (Entrez) 3310
Gene Symbol HSPA6
Host Species Human
Quantity 1 Kit
Storage Requirements Storage of components varies. See protocol for specific instructions.
Target HspA6
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.