Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human HTRA1/PRSS11 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 11 publications
Supplier: R&D Systems MAB2916SP
This item is not returnable.
View return policy
Description
HTRA1/PRSS11 Monoclonal specifically detects HTRA1/PRSS11 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
HTRA1/PRSS11 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
Q92743 | |
HTRA1 | |
Mouse myeloma cell line NS0-derived recombinant human HTRA1/PRSS11 Gln23-Pro480 Accession # Q92743 | |
25 μg | |
Primary | |
Detects human HTRA1/PRSS11 in direct ELISAs and Western blots. | |
Human | |
Purified |
Western Blot, Immunoprecipitation | |
275615 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ARMD7, EC 3.4.21, EC 3.4.21.108, High-temperature requirement A serine peptidase 1, HtrA serine peptidase 1, IGFBP5-protease, L56, ORF480, PRSS11, Serine protease 11, serine, 11 (IGF binding) | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5654 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction