Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human HTRA1/PRSS11 Antibody, R&D Systems™

Catalog No. AF2916
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2916 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF2916 Supplier R&D Systems Supplier No. AF2916
This item is not returnable. View return policy

Sheep Polyclonal Antibody

HTRA1/PRSS11 Polyclonal specifically detects HTRA1/PRSS11 in Human samples. It is validated for Western Blot, Simple Western.

Specifications

Antigen HTRA1/PRSS11
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Simple Western 10 ug/mL
Gene Accession No. Q92743
Gene Alias ARMD7, EC 3.4.21, EC 3.4.21.108, High-temperature requirement A serine peptidase 1, HtrA serine peptidase 1, IGFBP5-protease, L56, ORF480, PRSS11, Serine protease 11, serine, 11 (IGF binding)
Gene Symbols HTRA1
Host Species Sheep
Immunogen E. coli-derived recombinant human HTRA1/PRSS11 Gly156-Pro480 Accession # Q92743
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5654
Test Specificity Detects human HTRA1/PRSS11 in direct ELISAs and Western blots.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.