Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human HTRA2/Omi Quantikine ELISA Kit

Catalog No. DHTR20
Click to view available options
Quantity:
1 Kit
This item is not returnable. View return policy
This item is not returnable. View return policy

a 4.5 hour solid phase ELISA designed to measure HTRA2 levels in cell culture supernates, serum, and platelet-poor plasma.

The Human HTRA2/Omi Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human HTRA2/Omi in cell culture supernates (50 ul), serum (25 ul), platelet-poor edta plasma (25 ul), platelet-poor heparin plasma (25 ul).

Specifications

Assay Range 31.30 to 2000 pg/mL
Assay Sensitivity 13.2 pg/mL
Conjugate HRP
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Platelet-poor Plasma (EDTA, Heparin), Serum
Specificity Natural and recombinant HTRA2/Omi
For Use With (Application) ELISA
Gene ID (Entrez) 27429
Gene Symbol HTRA2
Quantity 1 Kit
Sample Volume Cell Culture Supernates (50uL),Serum (25uL),Platelet-poor EDTA Plasma (25uL),Platelet-poor Heparin Plasma (25uL)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym High temperature requirement protein A2, HtrA serine peptidase 2, HtrA2, HtrA-like serine protease, Omi, OMIOmi stress-regulated endoprotease, PARK13EC 3.4.21.108, PRSS25, PRSS25serine, 25, Serine protease 25, serine protease HTRA2, mitochondrial, Serine proteinase OMI
Target HTRA2/Omi
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.